Få mere at vide
MCT1/SLC16A1 Antibody (882616) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8275X
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
MCT1/SLC16A1 Monoclonal antibody specifically detects MCT1/SLC16A1 in Human samples. It is validated for Flow Cytometry, CyTOF
Tekniske data
| MCT1/SLC16A1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 882616 | |
| Flow Cytometry, CyTOF-ready | |
| FLJ36745, HHF7, MCT, MCT 1, MCT1MGC44475, member 1, monocarboxylate transporter 1, Solute carrier family 16 member 1, solute carrier family 16, member 1 (monocarboxylic acid transporter 1) | |
| NS0 mouse myeloma cell line transfected with human MCT1/SLC16A1, Met1-Val500, Accession # P53985 | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6566 | |
| Store at 4°C in the dark. | |
| IgG2a |