Få mere at vide
MedChemExpress MCLA hydrochloride
Tekniske data
Tekniske data
| Indhold og opbevaring | 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture) |
| Produkttype | Dye Reagents |
| Mængde | 5 mg |
| Forsendelsestilstand | Room temperature |
| CAS | 128322-44-1 |
| Molekylvægt (g/mol) | 291.73 g/mol |
| Opløselighed | DMSO : 10 mg/mL (34.28 mM; ultrasonic and warming and heat to 60°C) |
| Mål | NO Synthase ∣ MMP ∣ Endogenous Metabolite |