Få mere at vide
MCC-DM1, TRC
Artikelnummer. 30480122
Skift visning
Mængde:
0.5 mg
Pakningsstørrelse:
0.5mg
Artikelnummer. Mængde unitSize
30480122 0.5 mg 0.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30480122 Leverandør Toronto Research Chemicals Leverandørnr. TRCM1993000.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale MCC-DM1
CAS 1228105-53-0
Molekylær formel C47H63ClN4O14S
Mængde 0.5 mg
Synonym DM1-MCC, N2'-Deacetyl-N2'-[3-[[1-[(4-carboxycyclohexyl)methyl]-2,5-dioxo-3-pyrrolidinyl]thio]-1-oxopropyl]-maytansine
InChI formel InChI=1S/C47H63ClN4O14S/c1-25-10-9-11-35(63-8)47(61)23-33(64-45(60)49-47)26(2)41-46(4,66-41)36(22-38(54)51(6)31-19-29(18-25)20-32(62-7)40(31)48)65-44(59)27(3)50(5)37(53)16-17-67-34-21-39(55)52(42(34)56)24-28-12-14-30(15-13-28)43(57)58/h9-11,19-20,26-28,30,33-36,41,61H,12-18,21-24H2,1-8H3,(H,49,60)(H,57,58)/b11-9+,25-10+/t26-,27+,28?,30?,33+,34?,35-,36+,41-,46+,47+/m1/s1
SMIL O=C(C[C@H](OC([C@H](C)N(C)C(CCSC1C(N(CC2CCC(C(O)=O)CC2)C(C1)=O)=O)=O)=O)[C@@]([C@H]3[C@@H]4C)(O3)C)N(C)C5=CC(C/C(C)=C/C=C/[C@@H](OC)[C@]6(O)NC(O[C@H]4C6)=O)=CC(OC)=C5Cl
Molekylvægt (g/mol) 975.54
Formel vægt 974.375