Få mere at vide
MC-DOXHZN hydrochloride, MedChemExpress
Beskrivelse
MC-DOXHZN ((E/Z)-Aldoxorubicin) hydrochloride is an albumin-binding prodrug of Doxorubicin (DNA topoisomerase II inhibitor), with acid-sensitive properties.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | MC-DOXHZN hydrochloride |
| CAS | 480998-12-7 |
| Farve | Reddish Brown |
| Renhedsgrad noter | Research |
| Molekylær formel | C37H43ClN4O13 |
| Mængde | 5 mg |
| Synonym | (E/Z)-Aldoxorubicin hydrochlorideDoxorubicin(6-maleimidocaproyl)hydrazone hydrochloride |
| Opløselighedsinformation | DMSO : 25 mg/mL (31.76 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (15.88 mM; ultrasonic and warming and heat to 60°C) |
| SMIL | [H][C@@]1(O[C@H]2C[C@@](/C(CO)=N\NC(CCCCCN3C(C=CC3=O)=O)=O)(O)CC(C2=C4O)=C(O)C5=C4C(C6=C(OC)C=CC=C6C5=O)=O)O[C@@H](C)[C@@H](O)[C@@H](N)C1.[H]Cl |
| Molekylvægt (g/mol) | 787.21 |
| Vis mere |