missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
MBTD1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06299AF700
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
MBTD1 Polyclonal specifically detects MBTD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Tekniske data
| MBTD1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| mbt domain containing 1 | |
| Produced in rabbits immunized with E. coli-derived Human MBTD1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 54799 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion