missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody (HL1033), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-13687
Additional Details : Weight : 0.00970kg
Description
MBP Monoclonal antibody specifically detects MBP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
MBP | |
Monoclonal | |
Unconjugated | |
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of mouse MBP. The exact sequence is proprietary. | |
100 μg | |
Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers | |
4155 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
HL1033 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |