missing translation for 'onlineSavingsMsg'
Learn More

MBL™ Amber Soda-Lime Bottles

Ideal for the storage of light sensitive materials

Brand:  MBL SBA104

 View more versions of this product

Product Code. 15788047

  • 2145.00 DKK / Pack of 10
Only null left
Estimated Shipment: 21-10-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • Complies with ISO 4796-2
  • Amber soda-lime glass bottles with ground neck
  • UV absorbent up to 500nm
  • Supplied with polyethylene stoppers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

250 mL
69mm
10/Pk.
131 mm
Amber
Soda Lime Glass
19/26
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MBL™ Amber Soda-Lime Bottles > Capacity: 250; Height: 131mm; Standard taper joint: 19/26

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.