missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Maxi Potassium channel alpha Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1570.00 DKK - 3680.00 DKK
Tekniske data
| Antigen | Maxi Potassium channel alpha |
|---|---|
| Applikationer | Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Form | Purified |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
18302014
|
Novus Biologicals
NBP3-03012-20UL |
20 μL |
1570.00 DKK
20µL |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
18320996
|
Novus Biologicals
NBP3-03012-100UL |
100 μL |
3680.00 DKK
100µL |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
Maxi Potassium channel alpha Polyclonal specifically detects Maxi Potassium channel alpha in Rat samples. It is validated for Western Blot.Tekniske data
| Maxi Potassium channel alpha | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| bA205K10.1, BK channel, BKCA alpha, BKCA alpha subunit, BKTM, calcium-activated potassium channel subunit alpha-1, DKFZp686K1437, hSlo, k(VCA)alpha, KCNMA, Maxi K channel, MaxiK, MGC71881, potassium large conductance calcium-activated channel, subfamily M, alphamember 1, SAKCA, Slo homolog, Slo1, SLO-ALPHA, Slowpoke homolog, stretch-activated Kca channel, subfamily M subunit alpha-1 | |
| A synthetic peptide corresponding to a sequence within amino acids 850-950 of human Maxi Potassium channel alpha (NP_001258447.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Potassium Channels | |
| PBS with 50% glycerol, pH7.3. | |
| 3778 | |
| IgG | |
| Affinity purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel