missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Max Antibody (103), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90153AF750
Additional Details : Weight : 0.00970kg
Description
Max Monoclonal specifically detects Max in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Max | |
Monoclonal | |
Alexa Fluor 750 | |
BHLHD4, bHLHd4MGC11225, bHLHd5, bHLHd6, bHLHd7, bHLHd8, Class D basic helix-loop-helix protein 4, helix-loop-helix zipper protein, MAX protein, MGC10775, MGC18164, MGC34679, MGC36767, MYC associated factor X, Myc-associated factor X, orf1, protein max | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Max (NP_002373; Met1-Ser160). | |
0.1 mL | |
Apoptosis, Phospho Specific | |
4149 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
103 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |