missing translation for 'onlineSavingsMsg'
Få mere at vide

MedChemExpress Mastoparan

Artikelnummer. 16461898
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
Pakningsstørrelse:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16461898 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16461898 Leverandør MedChemExpress Leverandørnr. HYP0246

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Mastoparan, a tetradecapeptide which is a component of wasp venom, stimulates release of prolactin from cultured rat anterior pituitary cells.

Tekniske data

Renhed 95.47%
CAS 72093-21-1
Indhold og opbevaring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Til brug med (applikation) Metabolism-protein/nucleotide metabolism
Molekylvægt (g/mol) 1478.91 g/mol
Produkttype Natural Products
Mængde 5 mg
Forsendelsestilstand Room temperature
Opløselighed H2O : 33.33 mg/mL (22.54 mM∣Need ultrasonic)
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.