Learn More
Masterflex™ I/P™ End Bell Assembly
Complete your pump head assembly with two end bell assemblies. MasterFlex™ I/P™ End Bell Assemblies are designed for use with Masterflex™ I/P™ 73 Standard Pump Heads.
1630.00 DKK - 2195.00 DKK
Specifications
For Use With | For Masterflex I/P 73 standard pump heads |
---|---|
Quantity | 1 Each |
Type | End Bell Assembly |
Product Code | Brand | Material | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | ||||||
11860090
|
Masterflex™
07019-78 |
Polycarbonate, Cold-rolled Steel |
1630.00 DKK
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||||
11786068
|
Masterflex™
07019-79 |
Polycarbonate, Stainless Steel |
2195.00 DKK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
Description
- Assemblies are made from high-performance Polyphenylene sulfide and stainless steel
Specifications
For Masterflex I/P 73 standard pump heads | |
1 Each | |
End Bell Assembly |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Masterflex™ I/P™ End Bell Assembly