Få mere at vide
Marbofloxacin-d8, TRC
Artikelnummer. 30480025
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30480025 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30480025 Leverandør Toronto Research Chemicals Leverandørnr. TRCM1970025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Marbofloxacin-d8
CAS 1185053-37-5
Renhedsgrad noter HPLC
Molekylær formel C17 2H8 H11 F N4 O4
Mængde 5 mg
Synonym 9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl-2,2,3,3,5,5,6,6-d8)-7-oxo-7H-pyrido[3,2,1-ij][4,1,2]benzoxadiazine-6-carboxylic acid, Marbofloxacin-D8 (piperazinyl-2,2,3,3,5,5,6,6-D8), Marbofloxacin-d8
InChI formel InChI=1S/C17H19FN4O4/c1-19-3-5-21(6-4-19)14-12(18)7-10-13-16(14)26-9-20(2)22(13)8-11(15(10)23)17(24)25/h7-8H,3-6,9H2,1-2H3,(H,24,25)/i3D2,4D2,5D2,6D2
SMIL [2H]C1([2H])N(C)C([2H])([2H])C([2H])([2H])N(c2c(F)cc3C(=O)C(=CN4N(C)COc2c34)C(=O)O)C1([2H])[2H]
Molekylvægt (g/mol) 370.4
Formel vægt 370.1892
Procent renhed >95