Få mere at vide
Tocris Bioscience™ Maraviroc
GREENER_CHOICE
Artikelnummer. 18084382
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18084382 10 mg
18094942 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18084382 Leverandør Tocris Bioscience™ Leverandørnr. 3756/10
Selective CCR5 antagonist
Selective CCR5 antagonist; displays potent anti-HIV-1 activity. Prevents the interaction of HIV-1 gp120 and CCR5 (IC50 = 6.4 nM), inhibiting HIV-1 entry. Exhibits antinociceptive effects in a rat model of neuropathic pain. Also inhibits CCL3 (MIP-1α) binding to CCR5. Orally bioavailable.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 376348-65-1
Molekylær formel C29H41F2N5O
Tekniske data
Kemisk navn eller materiale Maraviroc
Mængde 10 mg
Formel vægt 513.67
Renhed >99%
Molekylvægt (g/mol) 513.678