Få mere at vide
Maprotiline-d5 Hydrochloride, TRC
Artikelnummer. 30480737
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30480737 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30480737 Leverandør Toronto Research Chemicals Leverandørnr. TRCM1880021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Maprotiline-d5 Hydrochloride
CAS 1794942-12-3
Molekylær formel C20H19D5ClN
Mængde 1 mg
Synonym N-Methyl-9,10-ethanoanthracene-9(10H)-propanamine-d5 Hydrochloride, N-Methyl-9,10-ethanoanthracene-9(10H)-propylamine-d5 Hydrochloride, Ba-34276-d5, Deprilept-d5, Ludiomil-d5, Psymion-d5
InChI formel InChI=1S/C20H23N.ClH/c1-21-14-6-12-20-13-11-15(16-7-2-4-9-18(16)20)17-8-3-5-10-19(17)20;/h2-5,7-10,15,21H,6,11-14H2,1H3;1H/i1D3,14D2;
SMIL [2H]C(NC([2H])([2H])[2H])([2H])CCC1(CC2)C3=C(C=CC=C3)C2C4=CC=CC=C41.Cl
Molekylvægt (g/mol) 318.9