missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAPK3 (ERK1) Recombinant Human Protein
MAPK3 (ERK1) is a serine/threonine kinase.
Brand: Invitrogen™ PR5254B
Additional Details : Weight : 0.01000kg
Description
MAPK3 (ERK1) is an important activator of p90, RSK, MSK, ELK1, and Stat3, and is involved in the MAPK pathway.
Specifications
NP_002737 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
MAPK3 (ERK1) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
70.8kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
MAP Kinase Family | |
Recombinant | |
E. coli |