missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAPK14 (p38 α) Recombinant Human Protein
MAPK14 (p38 alpha) is a member of the stress-activated protein kinase class of MAPKs.
Brand: Invitrogen™ PR5049C
Additional Details : Weight : 0.01000kg
Specifications
NP_620581 | |
20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
MAPK14 (p38 α) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
67.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
MAP Kinase Family | |
Recombinant | |
E. coli |