missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MAPK13 Recombinant Protein
Brand: Abnova™ H00005603-P01.25ug
This item is not returnable.
View return policy
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Antibody Production, ELISA, Protein Array, Western Blot | |
65.78 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 μg | |
-80°C | |
Recombinant |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
Human MAPK13 Full-length ORF Recombinant Protein with GST-tag at N-terminal | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE stained with Coomassie Blue | |
Wheat Germ (in vitro) | |
Human | |
Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ MAPK13 Recombinant Protein > 25μg
Spot an opportunity for improvement?Share a Content Correction