Få mere at vide
Mapenterol-d6 Hydrochloride, TRC
Artikelnummer. 30469514
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30469514 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469514 Leverandør Toronto Research Chemicals Leverandørnr. TRCM1870021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Mapenterol-d6 Hydrochloride
CAS 1246816-02-3
Molekylær formel C14H14D6ClF3N2O • HCl
Mængde 1 mg
Synonym 4-Amino-3-chloro-α-[[(1,1-dimethylpropyl-d6)amino]methyl]-5-(trifluoromethyl)benzenemethanol Hydrochloride
InChI formel InChI=1S/C14H20ClF3N2O.ClH/c1-4-13(2,3)20-7-11(21)8-5-9(14(16,17)18)12(19)10(15)6-8;/h5-6,11,20-21H,4,7,19H2,1-3H3;1H/i2D3,3D3;
SMIL NC1=C(C(F)(F)F)C=C(C(O)CNC(C([2H])([2H])[2H])(C([2H])([2H])[2H])CC)C=C1Cl.Cl
Molekylvægt (g/mol) 330.81