missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP4K5 (KHS1) Recombinant Human Protein
MAP4K5 (KHS1) is a serine/threonine kinase that belongs to the GCK family of kinases.
21190.00 DKK - 100400.00 DKK
Specifications
Accession Number | NP_942089 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 122.5kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468483
|
Invitrogen™
PR6671A |
100μg |
21190.00 DKK
100µg |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10610205
|
Invitrogen™
PV4014 |
1mg |
100400.00 DKK
1mg |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MAP4K5 (KHS1) has been implicated as an upstream regulator of MAP kinase signaling pathways.
Specifications
NP_942089 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
STE20 Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
122.5kDa | |
RUO - research use only | |
MAP4K5 (KHS1) | |
Human | |
GST Tag | |
Frozen |