missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAP4K4 (HGK), GST Tag Recombinant Protein
Human MAP4K4 (HGK), GST Tag Recombinant Protein
Brand: Invitrogen™ PV4013
Additional Details : Weight : 0.00900kg
Description
MAP4K4 (HGK) plays an important role in stress-activated MAPK core signaling pathways.
Specifications
NP_004825 | |
1mg | |
STE20 Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
65 kDa | |
MAP4K4 (HGK) | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: STE Group | |
Baculovirus | |
Recombinant, Catalytic Domain | |
Serine/Threonine Kinases |