missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAP4K4 (HGK), GST Tag Recombinant Protein
Human MAP4K4 (HGK), GST Tag Recombinant Protein
13290.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_004825 |
---|---|
Species | Human |
Recombinant | Recombinant |
Protein Tag | GST Tag |
Kinase Group | Ser⁄Thr Kinases: STE Group |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10250823
|
Invitrogen™
PV4013 |
1mg |
57500.00 DKK
1mg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15438503
|
Invitrogen™
PR7010B |
100μg |
13290.00 DKK
100µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MAP4K4 (HGK) plays an important role in stress-activated MAPK core signaling pathways.
Specifications
NP_004825 | |
Recombinant | |
Ser⁄Thr Kinases: STE Group | |
Kinases & Inhibitors | |
Liquid |
Human | |
GST Tag | |
Dry Ice | |
Recombinant, Catalytic Domain | |
Serine/Threonine Kinases |