missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
MAP4K3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-83181-0.1ML
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
MAP4K3 Polyclonal specifically detects MAP4K3 in Human samples. It is validated for Western Blot.
Tekniske data
| MAP4K3 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| EC 2.7.11, EC 2.7.11.1, germinal center kinase-like kinase, Germinal center kinase-related protein kinase, GLKMEKKK 3, MAPK/ERK kinase kinase kinase 3, MAPKKKK3, MEK kinase kinase 3, mitogen-activated protein kinase kinase kinase kinase 3, RAB8IPL1 | |
| The immunogen is a synthetic peptide directed towards the middle region of human MAP4K3 | |
| 0.1 mL | |
| Lipid and Metabolism, Protein Kinase | |
| 8491 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion