missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAP4K3 (GLK), GST Tag Recombinant Protein
Human MAP4K3 (GLK), GST Tag Recombinant Protein
3610.00 DKK - 54900.00 DKK
Specifications
Gene Alias | MAP4K3, MAPKKKK3, MEKKK3, RAB8IPL1 |
---|---|
Protein Length | aa 1-380 |
Species | Human |
Recombinant | Recombinant |
Protein Tag | GST Tag |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15458983
|
Invitrogen™
PV6349 |
10μg |
3610.00 DKK
10µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15478983
|
Invitrogen™
PV6351 |
1mg |
54900.00 DKK
1mg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15468983
|
Invitrogen™
PV6350 |
100 μg |
12310.00 DKK
100µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MAP4K3 is upstream of the MEKK1 target and is stimulated by UV radiation and TNF-alpha and can lead to the activation of the JNK signaling pathway. MAP4K3 can directly activate PKC-Θ, during TCR signaling and people with systemic lupus erythematosus show considerable enhanced MAP4K3 expression and activation of PKC-Θ, and the kinase IKK in T cells. MAP4K3 can act as a pro-apoptotic kinase which orchestrates activation of BAX via the concerted posttranscriptional modulation of PUMA, BAD, and BIM.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
MAP4K3, MAPKKKK3, MEKKK3, RAB8IPL1 | |
Human | |
GST Tag |
aa 1-380 | |
Recombinant |