missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP3K9 (MLK1) Recombinant Human Protein
MAP3K9 (MLK1) is a serine/threonine protein kinase.
13300.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_149132 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 83.4kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15428503
|
Invitrogen™
PR7007A |
100μg |
13300.00 DKK
100µg |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10118252
|
Invitrogen™
PV4012 |
1mg |
57500.00 DKK
1mg |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MAP3K9 (MLK1) functions in the MAP kinase pathway.
Specifications
NP_149132 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
MLK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
83.4kDa | |
RUO - research use only | |
MAP3K9 (MLK1) | |
Human | |
GST Tag | |
Frozen |