missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Manual Crimpers
Achieve secure, repeatable crimps or remove seals with minimized risk of damage with Thermo Scientific manual crimpers.
1645.00 DKK - 1870.00 DKK
Specifications
Type | Manual Crimper |
---|---|
For Use With (Equipment) | Autosampler Vial Closures |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
12490078
|
Thermo Scientific™
C4012-103 |
11mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12410268
|
Thermo Scientific™
C4020-103 |
20mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Lightweight, precision crimpers attach seals to chromatography or serum vials.
- Available for standard aluminum or flip-off seals.
- Factory set to properly crimp standard-thickness seals.
- Adjustable handle setting screw allows proper crimps with septa of varying thickness.
Specifications
Manual Crimper | |
Autosampler Vial Closures |