Få mere at vide
Manidipine dihydrochloride, MedChemExpress
Artikelnummer. 16457286
Encompass_Preferred
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16457286 100 mg
16443337 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16457286 Leverandør Medchem Express Leverandørnr. HY17403
MedChemExpress Manidipine dihydrochloride (CV-4093) is a dihydropyridine compound and a calcium channel blocker for Ca2+ current with IC50 of 2.6 nM.
Manidipine dihydrochloride (CV-4093) is a dihydropyridine compound and a calcium channel blocker for Ca2+ current with IC50 of 2.6 nM.
Tekniske data
Kemisk navn eller materiale Manidipine dihydrochloride
CAS 89226-75-5
Farve White
Renhedsgrad noter Research
Molekylær formel C35H40Cl2N4O6
Mængde 100 mg
Synonym CV-4093
Opløselighedsinformation DMSO : 50 mg/mL (73.14 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMIL O=C(C1=C(C)NC(C)=C(C(OC)=O)C1C2=CC=CC([N+]([O-])=O)=C2)OCCN3CCN(C(C4=CC=CC=C4)C5=CC=CC=C5)CC3.[H]Cl.[H]Cl
Molekylvægt (g/mol) 683.62
Formel vægt 683.62
Procent renhed 98.87%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation