missing translation for 'onlineSavingsMsg'
Learn More

Manganese Solution for ICP, 10g/L, HCl Matrix, Fisher Chemical™

Catalog Number p-8014477
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 10g/L
Quantity 100 mL
Solution Contains HCl
compliance-icons
Product Identifier
  • Single-element Standard Solution for ICP. Manganese (Mn) 10000mg/l in HCl 5% (S 233)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'