missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) Chloride Monohydrate, TRC

Product Code. 30470209
Change view
Click to view available options
Mængde:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30470209 50 g
30478920 5 g
30470399 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30470209 Supplier Toronto Research Chemicals Supplier No. TRCM16431850G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64333-01-3
Molekylær formel Cl2Mn . H2O
Molekylvægt (g/mol) 143.85
Synonym Manganese Chloride Monohydrate, Manganese dichloride monohydrate
IUPAC navn manganese(2+);dichloride;hydrate
SMIL O.[Cl-].[Cl-].[Mn+2]

Specifications

Kemisk navn eller materiale Manganese(II) Chloride Monohydrate
Mængde 50 g
InChI formel InChI=1S/2ClH.Mn.H2O/h2*1H;;1H2/q;;+2;/p-2
Formel vægt 142.8863
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.