Få mere at vide
MANF Antibody (012), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90750PCP
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
MANF Monoclonal antibody specifically detects MANF in Mouse samples. It is validated for ELISA
Tekniske data
| MANF | |
| 012 | |
| Protein A purified | |
| Purified |
| Monoclonal | |
| PBS | |
| RUO |