Få mere at vide
Maneb (>90%), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Maneb |
| CAS | 12427-38-2 |
| Molekylær formel | C4 H6 Mn N2 S4 |
| Mængde | 250 mg |
| Synonym | Manganese, [[1,2-ethanediylbis[carbamodithioato]](2-)]-, Manganese, [[2-[(dithiocarboxy)amino]ethyl]carbamodithioato(2-)-κS,κS']- (9CI), Manganese, [ethylenebis[dithiocarbamato]]- (8CI), Carbamodithioic acid, 1,2-ethanediylbis-, manganese complex, [N-[2-[(Dithiocarboxy)amino]ethyl]carbamodithioato(2-)-κS,κS']manganese, 1,2-Ethylenediylbis(carbamodithioato)manganese, AAmangan, Agrox N-M, Agrox flowable, Carbamodithioic acid, 1,2-ethanediylbis-, manganese(2+) salt (1:1), Dithane M 22, Ethylenebis[dithiocarbamic acid] manganous salt, F 10, F 10 (pesticide), Graneor, Maneb, Maneb 80, Manebgan, Manesan, Manex, Manganous ethylenebis[dithiocarbamate], Manzate, Manzate D, NM Drill Box, Nereb, Nespor, Plantifog 160M, Polyram M, Rhodianebe, Saneb M 22, Trimangol, Trimangol 80, Tritogol M, ZZ-Maneb Col |
| InChI formel | InChI=1S/C4H8N2S4.Mn/c7-3(8)5-1-2-6-4(9)10;/h1-2H2,(H2,5,7,8)(H2,6,9,10);/q;+2/p-2 |
| SMIL | S=C1NCCNC(=S)S[Mn]S1 |
| IUPAC navn | 1,3-dithia-5,8-diaza-2λ^{2}-manganacyclononane-4,9-dithione |
| Molekylvægt (g/mol) | 265.3 |
| Formel vægt | 264.8794 |