Få mere at vide
Emma Safety Footwear Mammoet M1807 XD Safety Shoes
Størrelse:
37
38
39
40
41
42
43
44
45
46
47
48
49
Pakningsstørrelse:
1 par
Beskrivelse
- S3-Safety shoe with sole plate for wet work
- GripForce™ Duo Heat Resistant Outsole
- Heat and cold insulated
- Fire-retardant laces
- Waterproof Oil Nubuck construction
- Hydro-Tec™ Sanitized Silver
Tekniske data
Tekniske data
| Certificeringer/compliance | EN ISO 20345:2011 |
| Lukketype | Laces |
| Farve | Black |
| Design | Mammoet M1807 XD |
| Liner materiale | Hydro-Tec™ Sanitized Silver / Sympatex™ |
| Materiale | Oiled Nubuck |
| Størrelse | 41 |
| Tå materiale | Steel |