Få mere at vide
Malvidin 3-O-Galactoside, TRC
Artikelnummer. 30483058
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483058 1 mg
30484036 5 mg
30483228 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30483058 Leverandør Toronto Research Chemicals Leverandørnr. TRCM1050151MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 30113-37-2
Molekylær formel C23 H25 O12 . Cl
Molekylvægt (g/mol) 528.89
Synonym 1-Benzopyrylium, 3-(β-D-galactopyranosyloxy)-5,7-dihydroxy-2-(4-hydroxy-3,5-dimethoxyphenyl)-, chloride (1:1), 1-Benzopyrylium, 3-(β-D-galactopyranosyloxy)-5,7-dihydroxy-2-(4-hydroxy-3,5-dimethoxyphenyl)-, chloride (9CI), Flavylium, 3-(galactosyloxy)-4',5,7-trihydroxy-3',5'-dimethoxy-, chloride (8CI), Primulin (6CI,7CI), Arthanitin, Malvidin 3-O-galactoside, Malvidin 3-O-β-D-galactopyranoside, Malvidin 3-galactoside, Malvidin galactoside
IUPAC navn (2S,3R,4S,5R,6R)-2-[5,7-dihydroxy-2-(4-hydroxy-3,5-dimethoxyphenyl)chromenylium-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol;chloride
SMIL [Cl-].COc1cc(cc(OC)c1O)c2[o+]c3cc(O)cc(O)c3cc2O[C@@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O
Tekniske data
Kemisk navn eller materiale Malvidin-3-galactoside chloride
Mængde 1 mg
InChI formel InChI=1S/C23H24O12.ClH/c1-31-14-3-9(4-15(32-2)18(14)27)22-16(7-11-12(26)5-10(25)6-13(11)33-22)34-23-21(30)20(29)19(28)17(8-24)35-23;/h3-7,17,19-21,23-24,28-30H,8H2,1-2H3,(H2-,25,26,27);1H/t17-,19+,20+,21-,23-;/m1./s1
Formel vægt 528.1035