missing translation for 'onlineSavingsMsg'
Learn More

Maltose Monohydrate, Technical, SLR, Fisher Chemical™

Catalog Number p-7073286
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
69-79-4
C12H22O11
342.30
MFCD00082026,MFCD00132834,MFCD18804057
GUBGYTABKSRVRQ-UHFFFAOYNA-N
maltose
134129532
(2R,3S,4S,5S,6R)-2-(hydroxymethyl)-6-[(2R,3R,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
This item is not returnable. View return policy
69-79-4
C12H22O11
342.30
MFCD00082026,MFCD00132834,MFCD18804057
GUBGYTABKSRVRQ-UHFFFAOYNA-N
maltose
134129532
(2R,3S,4S,5S,6R)-2-(hydroxymethyl)-6-[(2R,3R,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Maltose Monohydrate, Technical, SLR

TRUSTED_SUSTAINABILITY
Melting Point 130°C
Color White
Packaging Plastic powder jar
Quantity 100 g
Formula Weight 360.29g/mol
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'