missing translation for 'onlineSavingsMsg'
Få mere at vide

Maltooctaose, TRC

Artikelnummer. 30470818
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30470818 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30470818 Leverandør Toronto Research Chemicals Leverandørnr. TRCM16015025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Maltooctaose
CAS 6156-84-9
Molekylær formel C48 H82 O41
Mængde 25 mg
Synonym O-α-D-Glucopyranosyl-(1-4)-O-α-D-glucopyranosyl-(1-4)-O-α-D-glucopyranosyl-(1-4)-O-α-D-glucopyranosyl-(1-4)-O-α-D-glucopyranosyl-(1-4)-O-α-D-glucopyranosyl-(1-4)-O-α-D-glucopyranosyl-(1-6)-D-glucose
InChI formel InChI=1S/C48H82O41/c49-1-9-17(57)18(58)27(67)42(76-9)84-35-11(3-51)78-44(29(69)20(35)60)86-37-13(5-53)80-46(31(71)22(37)62)88-39-15(7-55)82-48(33(73)24(39)64)89-40-16(8-56)81-47(32(72)25(40)65)87-38-14(6-54)79-45(30(70)23(38)63)85-36-12(4-52)77-43(28(68)21(36)61)83-34-10(2-50)75-41(74)26(66)19(34)59/h9-74H,1-8H2/t9-,10-,11-,12-,13-,14-,15-,16-,17-,18+,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41?,42-,43-,44-,45-,46-,47-,48-/m1/s1
SMIL OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O[C@H]3[C@H](O)[C@@H](O)[C@@H](O[C@H]4[C@H](O)[C@@H](O)[C@@H](O[C@H]5[C@H](O)[C@@H](O)[C@@H](O[C@H]6[C@H](O)[C@@H](O)[C@@H](O[C@H]7[C@H](O)[C@@H](O)[C@@H](O[C@H]8[C@H](O)[C@@H](O)C(O)O[C@@H]8CO)O[C@@H]7CO)O[C@@H]6CO)O[C@@H]5CO)O[C@@H]4CO)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC navn (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-[(2R,3S,4R,5R,6R)-6-[(2R,3S,4R,5R,6R)-6-[(2R,3S,4R,5R,6R)-6-[(2R,3S,4R,5R,6R)-6-[(2R,3S,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molekylvægt (g/mol) 1315.14
Formel vægt 1314.4332
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.