Få mere at vide
Maleic Acid Monomethyl Ester (~90%), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Maleic Acid Monomethyl Ester |
| CAS | 3052-50-4 |
| Molekylær formel | C5H6O4 |
| Mængde | 50 g |
| Synonym | (2Z)-2-Butenedioic Acid 1-Methyl Ester, (2Z)-2-Butenedioic Acid Monomethyl Ester, Maleic Acid Methyl Ester, (Z)-4-Methoxy-4-oxobut-2-enoic Acid, (Z)-4-Methoxy-4-oxobut-2-enoic Acid, Hydrogen Methyl Maleate, Methyl Hydrogen (2Z)-2-Butendioate, Methyl Hydrogen Maleate, Monomethyl (Z)-2-Butenedioate, Monomethyl Maleate |
| InChI formel | InChI=1S/C5H6O4/c1-9-5(8)3-2-4(6)7/h2-3H,1H3,(H,6,7)/b3-2- |
| SMIL | COC(=O)\C=C/C(=O)O |
| IUPAC navn | (Z)-4-methoxy-4-oxobut-2-enoic acid |
| Molekylvægt (g/mol) | 130.1 |
| Formel vægt | 130.0266 |