Få mere at vide
Maleic-2,3-d2 Acid (contains ~1% d0), TRC
Artikelnummer. 30481730
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481730 100 mg
30466788 500 mg
30469435 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481730 Leverandør Toronto Research Chemicals Leverandørnr. TRCM124302100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 24461-33-4
Molekylær formel C4 D2 H2 O4
Molekylvægt (g/mol) 118.0845
Synonym 2-Butenedioic-2,3-d2 acid, (Z)- (9CI), Maleic-2,3-d2 acid (6CI, 8CI), Maleic-d2 acid, Maleic acid-2,3-D2, Maleic acid-d2, (Z)-2-Butenedioic-2,3-d2 acid, (2Z)-2-Butenedioic-2,3-d2 acid
IUPAC navn (Z)-2,3-dideuteriobut-2-enedioic acid
SMIL [2H]\C(=C(/[2H])\C(=O)O)\C(=O)O
Tekniske data
Kemisk navn eller materiale Maleic-2,3-d2 Acid
Mængde 100 mg
InChI formel InChI=1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1-/i1D,2D
Formel vægt 118.0235