missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Malathion, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Malathion |
| CAS | 121-75-5 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C10 H19 O6 P S2 |
| Mængde | 1 g |
| Synonym | Butanedioic acid, [(dimethoxyphosphinothioyl)thio]-, diethyl ester (9CI), Succinic acid, mercapto-, diethyl ester, S-ester with O,O-dimethyl phosphorodithioate (6CI,8CI), 8059HC, American Cyanamid 4,049, Atrapa, Carbetovur, Carbetox, Carbofos, Carbophos, Cimexan, Compound 4049, Cythion, Derbac M, Diethyl mercaptosuccinate S-ester with O,O-dimethyl phosphorodithioate, ENT 17,034, ENT 17034, Ethiolacar, Etiol, Extermathion, Forthion, Fosfothion, Fosfotion, Fyfanon, Gammaxine, Hi-Yield, Hilthion, Hymal, IFO 13140, Insecticide 4049, Insecticide no. 4049, Karbofos, Kunamala, Malafor, Malamar, Malamar 50, Malasol, Malaspray, Malataf, Malataf 57EC, Malathine, Malathion, Malathion E 50, Malathion LV Concentrate, Malathion ULV, Malathyl, Malathyne, Malatol, Malatol 500CE, Maldison, Mavidan, Mercaptothion, Moscarda, NSC 6524, Nassrlathion, O,O-Dimethyl S-(1,2-dicarbethoxyethyl) dithiophosphate, Oleophosphothion, Organoderm, Ortho Malathion, Ovide, Phosphothion, Prioderm, Radotion, Radotion E 50, Radotion P 5, S-[1,2-Bis(carbethoxy)ethyl] O,O-dimethyl dithiophosphate, S-[1,2-Bis(ethoxycarbonyl)ethyl] O,O-dimethyl phosphorodithioate, S-[1,2-Bis(ethoxycarbonyl)ethyl] O,O-dimethyl thiophosphate, SF 60, Sadofos, Sadophos, Security, Siptox I, Suleo M, Sumitox, TAK, TAK (pesticide), TM 4049, Vetiol, Zithiol, [(Dimethoxyphosphinothioyl)thio]butanedioic acid diethyl ester |
| InChI formel | InChI=1S/C10H19O6PS2/c1-5-15-9(11)7-8(10(12)16-6-2)19-17(18,13-3)14-4/h8H,5-7H2,1-4H3 |
| SMIL | CCOC(=O)CC(SP(=S)(OC)OC)C(=O)OCC |
| IUPAC navn | diethyl 2-dimethoxyphosphinothioylsulfanylbutanedioate |
| Molekylvægt (g/mol) | 330.36 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?