Få mere at vide
Malathion (Diethyl-d10), TRC
Artikelnummer. 30482621
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30482621 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482621 Leverandør Toronto Research Chemicals Leverandørnr. TRCM1110031MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Malathion D10 (diethyl D10)
CAS 347841-48-9
Molekylær formel C10 2H10 H9 O6 P S2
Mængde 1 mg
Synonym Malathion (diethyl-D10), 1,4-Di(ethyl-1,1,2,2,2-d5) 2-[(dimethoxyphosphinothioyl)thio]butanedioate, 2-[(Dimethoxyphosphinothioyl)thio]-,1,4-di(ethyl-1,1,2,2,2-d5)butanedioic Acid Ester
InChI formel InChI=1S/C10H19O6PS2/c1-5-15-9(11)7-8(10(12)16-6-2)19-17(18,13-3)14-4/h8H,5-7H2,1-4H3/i1D3,2D3,5D2,6D2
SMIL [2H]C([2H])([2H])C([2H])([2H])OC(=O)CC(SP(=S)(OC)OC)C(=O)OC([2H])([2H])C([2H])([2H])[2H]
IUPAC navn bis(1,1,2,2,2-pentadeuterioethyl) 2-dimethoxyphosphinothioylsulfanylbutanedioate
Molekylvægt (g/mol) 340.42
Formel vægt 340.0988