missing translation for 'onlineSavingsMsg'
Få mere at vide

Malathion-d7 (dimethyl-d6; 3-d1), CDN

Artikelnummer. 30320395
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320395 0.05 g
30310330 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320395 Leverandør CDN Leverandørnr. CDND51780.05G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Malathion D7 (dimethyl D6,3-D1)
CAS 352438-94-9
Molekylær formel C10 2H7 H12 O6 P S2
Mængde 0.05 g
Synonym Butanedioic-d acid, 3-[[di(methoxy-d3)phosphinothioyl]thio]-, diethyl ester (9 CI), Malathion D7 (dimethyl D6,3-D1)
InChI formel InChI=1 S/C10H19O6PS2/c1-5-15-9(11)7-8(10(12)16-6-2)19-17(18,13-3)14-4/h8H,5-7H2,1-4H3/i3D3,4D3,7 D
SMIL [2 H]C(C(SP(=S)(OC([2 H])([2 H])[2 H])OC([2 H])([2 H])[2 H])C(=O)OCC)C(=O)OCC
IUPAC navn diethyl 2-[bis(trideuteriomethoxy)phosphinothioylsulfanyl]-3-deuteriobutanedioate
Molekylvægt (g/mol) 337.4
Formel vægt 337.08 g/mol
Procent renhed 99 atom % D, min 95% Chemical Purity
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.