Få mere at vide
Malathion-d10 (diethyl-d10), CDN
Artikelnummer. 30314442
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30314442 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314442 Leverandør CDN Leverandørnr. CDND38450.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Malathion D10 (diethyl D10)
CAS 347841-48-9
Molekylær formel C10 2H10 H9 O6 P S2
Mængde 0.01 g
Synonym Malathion (diethyl-D10)
InChI formel InChI=1 S/C10H19O6PS2/c1-5-15-9(11)7-8(10(12)16-6-2)19-17(18,13-3)14-4/h8H,5-7H2,1-4H3/i1D3,2D3,5D2,6D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])OC(=O)CC(SP(=S)(OC)OC)C(=O)OC([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn bis(1,1,2,2,2-pentadeuterioethyl) 2-dimethoxyphosphinothioylsulfanylbutanedioate
Molekylvægt (g/mol) 340.42
Formel vægt 340.0988 g/mol
Procent renhed 99 atom % D, min 95% Chemical Purity