missing translation for 'onlineSavingsMsg'
Få mere at vide

Malathion 1000 μg/mL in Acetone, Dr. Ehrenstorfer

Artikelnummer. 30308971
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30308971 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30308971 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA14710000AC1000

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Malathion
CAS 121-75-5
Analyt- eller komponentnavne Malathion
Forsendelsestilstand Room Temperature
Molekylær formel C10 H19 O6 P S2
Koncentration 1000 μg/mL
Certificeringer/compliance ISO 17034
Synonym Butanedioic acid, 2-[(dimethoxyphosphinothioyl)thio]-, 1,4-diethyl ester, Butanedioic acid, [(dimethoxyphosphinothioyl)thio]-, diethyl ester (9CI), Succinic acid, mercapto-, diethyl ester, S-ester with O,O-dimethyl phosphorodithioate (6CI,8CI), 8059HC, American Cyanamid 4,049, Atrapa, Carbetovur, Carbetox, Carbofos, Carbophos, Cimexan, Compound 4049, Cythion, Derbac M, Diethyl mercaptosuccinate S-ester with O,O-dimethyl phosphorodithioate, ENT 17,034, ENT 17034, Ethiolacar, Etiol, Extermathion, Forthion, Fosfothion, Fosfotion, Fyfanon, Gammaxine, Hi-Yield, Hilthion, IFO 13140, Insecticide 4049, Insecticide no. 4049, Karbofos, Kunamala, Malafor, Malamar, Malamar 50, Malasol, Malaspray, Malataf, Malathine, Malathion, Malathion E 50, Malathion LV Concentrate, Malathion ULV, Malathyl, Malathyne, Malatol, Malatol 500CE, Maldison, Mavidan, Mercaptothion, Moscarda, NSC 6524, Nassrlathion, O,O-Dimethyl S-(1,2-dicarbethoxyethyl) dithiophosphate, Oleophosphothion, Organoderm, Ortho Malathion, Ovide, Phosphothion, Prioderm, Radotion, Radotion E 50, Radotion P 5, S-[1,2-Bis(carbethoxy)ethyl] O,O-dimethyl dithiophosphate, S-[1,2-Bis(ethoxycarbonyl)ethyl] O,O-dimethyl phosphorodithioate, S-[1,2-Bis(ethoxycarbonyl)ethyl] O,O-dimethyl thiophosphate, SF 60, Sadofos, Sadophos, Security, Siptox I, Suleo M, Sumitox, TAK, TAK (pesticide), TM 4049, Vetiol, Zithiol, [(Dimethoxyphosphinothioyl)thio]butanedioic acid diethyl ester, Diethyl (2RS)-2-(dimethoxyphosphinodithioyl)butanedioate
InChI formel InChI=1S/C10H19O6PS2/c1-5-15-9(11)7-8(10(12)16-6-2)19-17(18,13-3)14-4/h8H,5-7H2,1-4H3
SMIL CCOC(=O)CC(SP(=S)(OC)OC)C(=O)OCC
IUPAC navn diethyl 2-dimethoxyphosphinothioylsulfanylbutanedioate
Opløsningsmiddel eller Matrix Acetone
Molekylvægt (g/mol) 330.36
Mængde 1 ml
Formel vægt 330.0361 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.