Learn More
Mal-amido-peg6-NHS, TRC
Product Code. 30469110
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30469110 10 mg 10mg
30470006 100 mg 100mg
30472245 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30469110 Supplier Toronto Research Chemicals Supplier No. TRCM11105310MG
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1137109-21-7
Molecular Formula C26H39N3O13
Synonym Maleimide-PEG6-succinimidyl Ester∣N-Hydroxysuccinimidyl-(polyethylene glycol) 6-Maleimide∣SM(PEG)6
Specifications
Certifications/Compliance ISO 9001
Molecular Weight (g/mol) 601.6
Shipping Condition Room Temperature
Quantity 10 mg