Få mere at vide
Magnolol, TRC
Artikelnummer. 30132467
Skift visning
Mængde:
50 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132467 50 mg
30003683 500 mg
30007977 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132467 Leverandør Toronto Research Chemicals Leverandørnr. TRCM11039050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 528-43-8
Molekylær formel C18 H18 O2
Molekylvægt (g/mol) 266.33
Synonym 5,5'-Di-2-propen-1-yl-[1,1'-biphenyl]-2,2'-diol, 2,2'-Bichavicol, 5,5'-Diallyl-2,2'-biphenyldiol, Magnolol, NSC 293099
IUPAC navn 2-(2-hydroxy-5-prop-2-enylphenyl)-4-prop-2-enylphenol
SMIL Oc1ccc(CC=C)cc1c2cc(CC=C)ccc2O
Tekniske data
Kemisk navn eller materiale Magnolol
Mængde 50 mg
InChI formel InChI=1S/C18H18O2/c1-3-5-13-7-9-17(19)15(11-13)16-12-14(6-4-2)8-10-18(16)20/h3-4,7-12,19-20H,1-2,5-6H2
Formel vægt 266.1307