Få mere at vide
Magnoflorine Hydroxide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Magnoflorine Hydroxide |
| CAS | 476-63-1 |
| Molekylær formel | C20H25NO5 |
| Mængde | 10 mg |
| Synonym | (6aS)-5,6,6a,7-Tetrahydro-1,11-dihydroxy-2,10-dimethoxy-6,6-dimethyl-4H-dibenzo[de,g]quinolinium Hydroxide, (+)-Magnoflorine Hydroxide, Escholin Hydroxide, Escholine Hydroxide, Magnoflorine Hydroxide, Thalictrin Hydroxide, Thalictrine Hydroxide, α-Magnoflorine Hydroxide |
| SMIL | [OH-].COc1ccc2C[C@H]3c4c(CC[N+]3(C)C)cc(OC)c(O)c4c2c1O |
| Molekylvægt (g/mol) | 359.42 |