missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Mouse CD4 T cell Enrichment Kit

Designed for the magnetic separation of mouse CD4 T cells by negative selection. It has been optimized for the isolation of mouse CD4 T cells from spleens or lymph nodes utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads.

Brand:  Invitrogen™ 8804-6821-74

3745.30 DKK valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 15571277

  • 5750.00 DKK / 200 tests

Please to purchase this item. Need a web account? Register with us today!

Alert:

Contains Sodium azide


Includes:
  • MagniSort™ Mouse CD4 T cell Enrichment Antibody Cocktail : 200 tests, 20ÎĽL/test
  • MagniSort™ Negative Selection Beads B
This item is not returnable. View return policy

Description

Description

Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD4 T cells untouched and free in solution.

The MagniSort™ Mouse CD4 T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Mouse CD8 Biotin
Anti-Mouse CD11b Biotin
Anti-Mouse CD19 Biotin
Anti-Mouse CD24 Biotin
Anti-Mouse CD45R (B220) Biotin
Anti-Mouse CD49b (Integrin alpha 2) Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse γδ TCR Biotin
Anti-Mouse TER-119 Biotin
TRUSTED_SUSTAINABILITY
Specifications

Specifications

T Cells (CD4)
200 tests
Mouse
Cell Enrichment Kit
200 tests
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2459625 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number8804-6821-74
Lot Number2472011 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number8804-6821-74
Lot Number2539121 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number8804-6821-74
Lot Number2536249 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number8804-6821-74
Lot Number2599251 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number8804-6821-74
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ MagniSort™ Mouse CD4 T cell Enrichment Kit > MagniSort Mouse CD4 T cell Enrichment Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.