Learn More
Invitrogen™ MagniSort™ Human CD8 Positive Selection Kit
Designed for the magnetic separation of CD8+ cells by positive selection. It has been optimized for the isolation of CD8+ cells from human peripheral blood monocuclear cells, utilizing a biotinylated Anti-Human CD8 antibody and streptavidin-coated magnetic beads.
Brand:  Invitrogen™ 8802-6832-74
Product Code. 15561247
  • 5960.00 DKK / 200 tests
Alert:
Contains Sodium azide
Includes:
  • MagniSort Anti-Human CD8 Biotin : 200 tests, 20ÎĽL/test
  • MagniSort Positive Selection Beads A
Description
Description
  • CD8+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD8+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Human CD8, clones RPA-T8 or SK1
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD8+
200 tests
Human
Positive Selection Kit
200 tests
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2992811 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog Number8802-6832-74
Lot Number2913936 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number8802-6832-74
Lot Number2528510 Certificate TypeCertificate of Analysis Date02/09/2022 Catalog Number8802-6832-74
Promotions
Promotions
Product Content Correction
Product Title
Invitrogen™ MagniSort™ Human CD8 Positive Selection Kit > MagniSort Human CD8 Positive Selection Kit