Learn More
Invitrogen™ MagniSort™ Human CD8 Naïve T cell Enrichment Kit
Designed for the magnetic separation of CD8 naïve T cells by negative selection. It has been optimized for the isolation of human CD8 naïve T cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads.
Brand: Invitrogen™ 8804-6816-74
4428.70 DKK valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Alert:
Contains Sodium azide
Includes:
- MagniSort Human CD8 Naïve T cell Enrichment Antibody Cocktail : 200 tests, 20μL/test
- MagniSort Negative Selection Beads A
Description
Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD8 naïve T cells untouched and free in solution.The MagniSort™ Human CD8 Naïve T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD4 Biotin
Anti-Human CD11b Biotin
Anti-Human CD14 Biotin
Anti-Human CD16 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD36 Biotin
Anti-Human CD45RO Biotin
Anti-Human CD56 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
Anti-Human γδ TCR Biotin

Specifications
T Cells (Naïve, CD4) | |
200 tests | |
Human |
Cell Enrichment Kit | |
200 tests |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2344863 | Certificate of Analysis | 31/01/2025 | 8804-6816-74 |
2992758 | Certificate of Analysis | 31/01/2025 | 8804-6816-74 |
2545633 | Certificate of Analysis | 29/01/2025 | 8804-6816-74 |
2747827 | Certificate of Analysis | 29/01/2025 | 8804-6816-74 |
2887080 | Certificate of Analysis | 26/01/2025 | 8804-6816-74 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.