Learn More
Vitlab™ Magnetic Stir Bar Retriever
Vitlab™ Magnetic Stir Bar Retriever is constructed of polyethylene and comes individually packaged in a polyethylene bag.
Brand:  Vitlab™ 318393
Additional Details : Weight : 0.04500kg
Product Code. 10102651
  • 213.00 DKK / Each
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • With a permanent magnet on one end and holding ring on the other one
  • Magnet is completely encapsulated in a PE mantle
Specifications
Specifications
Polyethylene
450 mm