Learn More
Vitlab™ Magnetic Stir Bar Retriever
Vitlab™ Magnetic Stir Bar Retriever is constructed of polyethylene and comes individually packaged in a polyethylene bag.
Brand: Vitlab™ 318393
Additional Details : Weight : 0.04500kg
Description
- With a permanent magnet on one end and holding ring on the other one
- Magnet is completely encapsulated in a PE mantle
Specifications
Polyethylene | |
450 mm |