missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium sulfate hydrate, Puriss., meets analytical spec. of DAC, dried, 99.0-101.0% MgSO4 basis (in dried substance), Honeywell Fluka™
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Physical Form | Dry |
Quantity | 1 kg |
Linear Formula | MgSO4 · aq |
UN Number | NONH for all modes of transport |
IUPAC Name | magnesium;sulfate |
Formula Weight | 120.37g/mol (anhydrous basis) |
Percent Purity | 99 to 101% (MgSO4 basis) (in dried substance) |
Grade | puriss. |
Chemical Name or Material | Magnesium Sulfate Hydrate, Meets analytical specification of DAC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'