missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate Hexahydrate, puriss. p.a., ≥99.0% (KT), Honeywell™ Fluka™

Brand:  Honeywell Fluka 00627-250G

Additional Details : CAS Number : 17830-18-1 Weight : 0.25000kg

Packaging Poly Bottle
Quantity 250g

 View more versions of this product

Product Code. 15640520

  • 189.03 DKK / 250g


Estimated Shipment date
from Supplier 25-07-2018
Add to basket

Specifications

Chemical Identifiers

Magnesium Sulfate Hexahydrate
puriss. p.a.
Colorless
228.46g/mol
5 to 8
NONH for all modes of transport
≥99.0% (KT)
Solid
Crystals
MgSO4 · 6H2O
MFCD00149786
17830-18-1

Specifications

Magnesium Sulfate Hexahydrate
puriss. p.a.
MgSO4 · 6H2O
228.46g/mol
NONH for all modes of transport
Solid
5 to 8
17830-18-1
≥99.0% (KT)
Crystals
MFCD00149786
Poly Bottle
Colorless
250g
Videos